Order Entry
Czech Republic
ContactUsLinkComponent
Rp-8-Br-cGMPS ≥98% (by HPLC)
Rp-8-Br-cGMPS ≥98% (by HPLC)
Catalog # ENZOBMLCN2160500
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Rp-8-Br-cGMPS ≥98% (by HPLC)
Catalog # ENZOBMLCN2160500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :462.2 g/mol
Storage Temperature:Freezer
CAS Number:150418-07-8
Formula:C₁₀H₁₀BrN₅O₆PS·Na