Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cdk/Crk inhibitor
Cdk/Crk inhibitor ≥98% (by HPLC)
Cdk/Crk inhibitor ≥98% (by HPLC)
Cdk/Crk inhibitor ≥98% (by HPLC)
Catalog # ENZOENZCHM1310001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Cdk/Crk inhibitor ≥98% (by HPLC)
Catalog # ENZOENZCHM1310001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
473.4 g/mol
CAS Number:
784211-09-2
Formula:
C₂₃H₂₂Cl₂N₄O₃
Product Details