BML-277 Chk2 inhibitor ≥98% (by TLC)
Catalog # ENZOBMLEI3880010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:10 mg
- Sekvence:2275
Specifications
About this item
:
:363.8 g/mol
Storage Temperature:Freezer
CAS Number:516480-79-8
Formula:C₂₀H₁₄ClN₃O₂
MDL Number:MFCD08276917