Order Entry
Czech Republic
ContactUsLinkComponent
3-O-Acetyl-11-keto-β-boswellic acid (from Boswellia serrata) ≥98% (by HPLC)
3-O-Acetyl-11-keto-β-boswellic acid (from Boswellia serrata) ≥98% (by HPLC)
Catalog # ENZOALX350310M005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
3-O-Acetyl-11-keto-β-boswellic acid (from Boswellia serrata) ≥98% (by HPLC)
Catalog # ENZOALX350310M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :512.7 g/mol
Storage Temperature:Freezer
CAS Number:67416-61-9
Formula:C₃₂H₄₈O₅
MDL Number:MFCD03788777