Order Entry
Czech Republic
ContactUsLinkComponent
FABP4 inhibitor ≥99% (by HPLC)
FABP4 inhibitor ≥99% (by HPLC)
Catalog # ENZOENZCHM1180005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
FABP4 inhibitor ≥99% (by HPLC)
Catalog # ENZOENZCHM1180005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :474.5500 g/mol
CAS Number:300657-03-8
Formula:C₃₁H₂₆N₂O₃
MDL Number:MFCD09991687