Order Entry
Czech Republic
ContactUsLinkComponent
10-DEBC hydrochloride ≥98% (by HPLC)
10-DEBC hydrochloride ≥98% (by HPLC)
Catalog # ENZOENZCHM1260001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
10-DEBC hydrochloride ≥98% (by HPLC)
Catalog # ENZOENZCHM1260001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :381.4 g/mol
CAS Number:201788-90-1