Order Entry
Czech Republic
ContactUsLinkComponent
Src inhibitor-1 ≥98% (by HPLC)
Src inhibitor-1 ≥98% (by HPLC)
Catalog # ENZOBMLEI3980025
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Src inhibitor-1 ≥98% (by HPLC)
Catalog # ENZOBMLEI3980025
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :373.41 g/mol
Storage Temperature:Ambient
CAS Number:179248-59-0
Formula:C₂₂H₁₉N₃O₃
MDL Number:MFCD01815300