Order Entry
Czech Republic
ContactUsLinkComponent
Sodium 4-phenylbutyrate ≥98% (by UHPLC)
Sodium 4-phenylbutyrate ≥98% (by UHPLC)
Catalog # ENZOBMLEI3200001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Sodium 4-phenylbutyrate ≥98% (by UHPLC)
Catalog # ENZOBMLEI3200001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.2 g/mol
Storage Temperature:Ambient
CAS Number:1716-12-7
Formula:C₁₀H₁₁O₂·Na
MDL Number:MFCD00800247