Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TAPI-1
TAPI-1 ≥98% (by HPLC)
TAPI-1 ≥98% (by HPLC)
Catalog # ENZOBMLPI1340001
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
TAPI-1 ≥98% (by HPLC)
Catalog # ENZOBMLPI1340001
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
CAS Number:
171235-71-5
Product Details