N,N-Dimethylsphingosine, D-erythro ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:119567-63-4
:
Frequently Bought Together
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
Media bottles, square, sterile, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Lids for 96-well microplates
Supplier: Greiner Bio-One
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Corning® Microplate Sealing Tapes
Supplier: Corning
Cryoboxes, 133×133 mm
Supplier: TENAK
Microplates, 96-well
Supplier: Greiner Bio-One
Inoculating loops, stainless steel
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Optical bottom plates, SCREENSTAR
Supplier: Greiner Bio-One
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific