Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
GSK-3β inhibitor XVIII
GSK-3β inhibitor XVIII ≥98% (by HPLC)
GSK-3β inhibitor XVIII ≥98% (by HPLC)
GSK-3β inhibitor XVIII ≥98% (by HPLC)
Catalog # ENZOENZCHM1340005
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
GSK-3β inhibitor XVIII ≥98% (by HPLC)
Catalog # ENZOENZCHM1340005
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
413.9 g/mol
CAS Number:
1139875-74-3
Product Details