
Guanylylimidodiphosphate, trisodium salt ≥95% (by HPLC)
Catalog # ENZOENZCHM1000005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:≥95% (HPLC)
Specification Test Results
About this item
:
:588.14 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:148892-91-5
Formula:C₁₀H₁₄N₆Na₃O₁₃P₃