Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
IDE 2 ≥98% (by TLC)

IDE 2 ≥98% (by TLC)

1 mg
Katalogové číslo ENZOBMLSC1010001
Dodavatel:   Enzo Life Sciences
IDE 2 ≥98% (by TLC)
Katalogové číslo ENZOBMLSC1010001
Dodavatel:  Enzo Life Sciences
Číslo CAS:  1136466-93-7
IDE 2 ≥98% (by TLC)
IDE 2 ≥98% (by TLC)

1 mg

Přejít na:
Číslo CAS:  1136466-93-7
Price
4 240,00 Kč
Přihlásit se pro dostupnost
1 * 1 mg
Product Details & Documents

Induces stem cell differentiation.

Cell-permeable molecule capable of directing differentiation of mouse and human embryonic stem cells into endodermal cells (EC₅₀ = 223 nM). Developmental potential of these chemically derived endodermal cells has been shown via continued differentiation into pancreatic progenitor cells and incorperation into developing gut tubes of live mouse embryos. Nodal/Smad signaling is activated indicating IDE-2 functions via TGF-β-signaling pathway.

Soluble in DMSO (>25 mg/ml) or 100% ethanol (25 mg/ml).

Synonyma:  IDE-2, 7-(2-Cyclopentylidenehydrazinyl)-7-oxoheptanoic acid
  • Číslo CAS:1136466-93-7
  • Vzorec:C₁₂H₂₀N₂O₃
  • Molecular Weight:240.3 g/mol
  • Teplota pro skladování:Freezer
Specifikace

Výsledky testů specifikací

  • Purity:
    ≥98% (TLC)

Specifikace

  • Pk:
    1 mg

Specifikace