
L-858051 ≥92% (by HPLC)
Catalog # ENZOBMLCN1050005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:≥92% (HPLC)
Specification Test Results
About this item
:
:627.6 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:115116-37-5
Formula:C₂₉H₄₈N₂O₇·2HCl·H₂O