Motesanib ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:453562-69-1
:
Formula:C₂₂H₂₃N₅O
MDL Number:MFCD10567689
:373.5 g/mol
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
VWR® Media Bottles, Square, PETG
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Dewar flasks, Nalgene®
Supplier: Thermo Fisher Scientific
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Bottles, PET, Square, with Screw Cap
Supplier: VWR Collection
Containers with optional snap caps, Nunc™
Supplier: Thermo Fisher Scientific
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Test tubes with screw cap
Supplier: KARTELL
Conical-bottom centrifuge tube adapters, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
J.T.Baker®, Plate 384
Supplier: Avantor
VWR®, Blotting Pads, Grade 707
Supplier: VWR Collection
BAGOPENER FOR STOMACHER 400 ECO BAGS 1 * 1 items
Supplier: Seward (FermionX)