Order Entry
Czech Republic
ContactUsLinkComponent
Zinquin free acid ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Zinquin free acid ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Catalog # ENZOENZ52152
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Zinquin free acid ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Catalog # ENZOENZ52152
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :386.42 g/mol
CAS Number:151606-29-0
Formula:C₁₉H₁₈N₂O₅S
MDL Number:MFCD03452711