HOE-140 ≥95% (by HPLC)
Catalog # ENZOBMLNK1040001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Purity:≥95% (HPLC)
- Identity:Determined by MS
- Sequence:D-Arg-Arg-Pro-Hyp-Gly-Thi-Ser-Tic-Oic-Arg
Specification Test Results
About this item
:
:1304.5 g/mol
Storage Temperature:Freezer
CAS Number:130308-48-4
Formula:C₅₉H₈₉N₁₉O₁₃S