Order Entry
Czech Republic
ContactUsLinkComponent
 
5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid ≥95% (by HPLC)
  ENZOBMLK0050050
 :  
undefined
5-Ketoeicosa-6E,8Z,11Z,14E-tetraenoic acid ≥95% (by HPLC)
  ENZOBMLK0050050
 :  

 

  • Pk:
    50 µG
  • Seq:
    1202

 

 

  • Concentration:
    50 µg/ml
  • Formulation:
    Oil dissolved in ethanol
  • Purity:
    ≥95% (HPLC)

 

 

 :
 :318.5 g/mol
 :Ultra-Low Temperature Freezer (–80°C)
 :1170
 :3,II
 :106154-18-1
 :C₂₀H₃₀O₃