
15(S)-Hydroxyeicosa-5Z,8Z,11Z,13E-tetraenoic acid (HETE) ≥99% (by HPLC)
:54845-95-3
:
:320.5 g/mol
:Ultra-Low Temperature Freezer (–80°C)
:1170
:3,II
:GHS02
- Source:Synthetic
- Formulation:Oil dissolved in ethanol
- Purity:≥99% (HPLC)
Frequently Bought Together