

TTNPB ≥98% (by HPLC)
Katalogové číslo ENZOBMLGR1050005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by H-NMR
- Appearance:White crystalline solid
Specification Test Results
About this item
:
:348.5 g/mol
:GHS07
:GHS08
CAS Number:71441-28-6
Formula:C₂₄H₂₈O₂
MDL Number:MFCD00673917