AMPA ≥98% (by TLC), white to off-white solid
Catalog # ENZOBMLEA1100005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Source:Synthetic
- Purity:≥98%
- Identity:Determined by NMR
Specification Test Results
About this item
:
:186.2 g/mol
Storage Temperature:Freezer
CAS Number:77521-29-0
Formula:C₇H₁₀N₂O₄