Order Entry
Czech Republic
ContactUsLinkComponent
5(S),15(S)-DIHETE ≥99% (by HPLC)
Flammable
5(S),15(S)-DIHETE ≥99% (by HPLC)
Catalog # ENZOBMLD5150050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Flammable
5(S),15(S)-DIHETE ≥99% (by HPLC)
Catalog # ENZOBMLD5150050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

Specification Test Results

  • Concentration:
    50 µg/ml
  • Formulation:
    Oil dissolved in ethanol
  • Purity:
    ≥99% (HPLC)

Specification Test Results

About this item

 :
 :336.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
CAS Number:82200-87-1
Formula:C₂₀H₃₂O₄