Order Entry
Czech Republic
ContactUsLinkComponent
trans-7-Hydroxy-PIPAT maleate ≥98%, white solid
trans-7-Hydroxy-PIPAT maleate ≥98%, white solid
Catalog # ENZOBMLD1060005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
trans-7-Hydroxy-PIPAT maleate ≥98%, white solid
Catalog # ENZOBMLD1060005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98%

Specification Test Results

About this item

 :
 :487.3 g/mol
Storage Temperature:Freezer
CAS Number:159559-71-4