CP-55,940 ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:83002-04-4
:
Formula:C₂₄H₄₀O₃
:376.6 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity:≥98% (HPLC)
- Identity:Determined by ¹H-NMR and MS
Specification Test Results
Frequently Bought Together