Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sp-8-Br-cGMPS
Sp-8-Br-cGMPS ≥98% (by HPLC)
Sp-8-Br-cGMPS ≥98% (by HPLC)
Sp-8-Br-cGMPS ≥98% (by HPLC)
ENZOBMLCN2170500
:
Enzo Life Sciences
:
Sp-8-Br-cGMPS ≥98% (by HPLC)
ENZOBMLCN2170500
:
Enzo Life Sciences
:
Pk:
500 µG
Seq:
1418
Purity:
≥98% (HPLC)
:
:
462.2 g/mol
:
Freezer
:
153660-03-8
:
C₁₀H₁₀BrN₅O₆PS·Na