Order Entry
Czech Republic
ContactUsLinkComponent
Sp-5,6-dichloro-cBIMPS ≥98% (by HPLC)
Sp-5,6-dichloro-cBIMPS ≥98% (by HPLC)
Catalog # ENZOBMLCN1200001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Sp-5,6-dichloro-cBIMPS ≥98% (by HPLC)
Catalog # ENZOBMLCN1200001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :419.2 g/mol
Storage Temperature:Freezer
CAS Number:120912-54-1
Formula:C₁₂H₁₀N₂O₅PSCl₂·Na