Fasudil dihydrochloride ≥98% (by TLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:203911-27-7
:103745-39-7
Formula:C₁₄H₁₇N₃O₂S·2HCl
MDL Number:MFCD00153805
:364.29 g/mol
Storage Temperature:Freezer
Frequently Bought Together