Order Entry
Czech Republic
ContactUsLinkComponent
Erbstatin analog ≥98% (by TLC)
Erbstatin analog ≥98% (by TLC)
Catalog # ENZOBMLEI1460005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Erbstatin analog ≥98% (by TLC)
Catalog # ENZOBMLEI1460005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :194.19 g/mol
CAS Number:63177-57-1
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00132932