

Dihydroergocristine methanesulphonate ≥98%, white solid
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:24730-10-7
:
Formula:C₃₆H₄₅N₅O₈S
MDL Number:MFCD00153792
:707.843 g/mol
UN:1544
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Purity:≥98%
Specification Test Results
Frequently Bought Together