Order Entry
Czech Republic
ContactUsLinkComponent
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] ≥95% (by HPLC), Ultrapure
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] ≥95% (by HPLC), Ultrapure
Katalogové číslo ENZOENZ52452
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
5(6)-Tetramethylrhodamine isothiocyanate [5(6)-TRITC] ≥95% (by HPLC), Ultrapure
Katalogové číslo ENZOENZ52452
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :443.52 g/mol
Storage Temperature:Freezer
CAS Number:95197-95-8
Formula:C₂₅H₂₁N₃O₃S
MDL Number:MFCD00063517