Troglitazone ≥98% (by TLC)
:97322-87-7
:
:C₂₄H₂₇NO₅S
:MFCD00878416
:441.5 g/mol
Frequently Bought Together
Sealing film, Axygen® AxySeal
: Corning
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
Multiwell cell culture plates, Falcon®
: Corning
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Serological pipettes, Stripette™
: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection