Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
17-AAG
17-AAG ≥98% (by TLC)
17-AAG ≥98% (by TLC)
17-AAG ≥98% (by TLC)
Catalog # ENZOBMLEI3080001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
17-AAG ≥98% (by TLC)
Catalog # ENZOBMLEI3080001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
585.7 g/mol
CAS Number:
75747-14-7
Formula:
C₃₁H₄₃N₃O₈
MDL Number:
MFCD04973892
Product Details