Order Entry
Czech Republic
ContactUsLinkComponent
 
4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione ≥95% (by TLC)
Catalog # ENZOBMLEI3990010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione ≥95% (by TLC)
Catalog # ENZOBMLEI3990010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :283.93 g/mol
Storage Temperature:Ambient
CAS Number:30675-13-9
Formula:C₉H₂Cl₄O₂
MDL Number:MFCD00239209