N-(2-Hydroxyethyl)oleamide ≥98% (by TLC)
Catalog # ENZOBMLSL2310050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:325.54 g/mol
Storage Temperature:Freezer
CAS Number:111-58-0
Formula:C₂₀H₃₉NO₂
MDL Number:MFCD00045972