Order Entry
Czech Republic
ContactUsLinkComponent
2,3-Diphenylquinoxaline-6-carboxylic acid ≥98% (by TLC)
2,3-Diphenylquinoxaline-6-carboxylic acid ≥98% (by TLC)
Catalog # ENZOBMLEI3360010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
2,3-Diphenylquinoxaline-6-carboxylic acid ≥98% (by TLC)
Catalog # ENZOBMLEI3360010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :326.36 g/mol
Storage Temperature:Ambient
CAS Number:32387-96-5
Formula:C₂₁H₁₄N₂O₂
MDL Number:MFCD00555042