15-Deoxy-δ-12,14-prostaglandin J2 ≥97% (by TLC)
:87893-55-8
:
:C₂₀H₂₈O₃
:MFCD00065806
:316.441 g/mol
:Freezer
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR®, Petri Dish, PS
: VWR Collection
Centrifuge tubes, 15 ml, Corning®
: Corning
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
Cell scrapers and cell lifters
: VWR Collection
VIAL 124ML HIGH PROFILE CLEAR N/L M800 1 * 250 items
: Capitol Europe
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
Snap cap for round bottom tubes, Falcon®
: Corning
VWR®, Square Bottles, PET
: LAB3 OWN PRODUCTION
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
VWR®, Cryoboxes, Half Size, 136×66,5 mm
: VWR Collection
Cell culture flasks with standard or filter caps, CELLSTAR®
: Greiner Bio-One
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR®, Sample Dippers
: VWR Collection
Multiwell cell culture plates, Falcon®
: Corning
Storage tubes, 2D barcoded, Matrix™
: THERMO MATRIX TECHNOLOGIES