Order Entry
Czech Republic
ContactUsLinkComponent
 
D-erythro-C8-ceramine ≥98% (by TLC)
Catalog # ENZOBMLSL1450005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
D-erythro-C8-ceramine ≥98% (by TLC)
Catalog # ENZOBMLSL1450005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :411.715 g/mol
Storage Temperature:Freezer
CAS Number:170926-06-4
Formula:C₂₆H₅₃NO₂
MDL Number:MFCD01632747