Order Entry
Czech Republic
ContactUsLinkComponent
N-Arachidonyl-2-chloroethylamide ≥98% (by TLC)
N-Arachidonyl-2-chloroethylamide ≥98% (by TLC)
Catalog # ENZOBMLFA0320050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
N-Arachidonyl-2-chloroethylamide ≥98% (by TLC)
Catalog # ENZOBMLFA0320050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)
  • Appearance:
    Pale yellow oil

Specification Test Results

About this item

 :
 :384.001 g/mol
Storage Temperature:Freezer
CAS Number:220556-69-4
Formula:C₂₂H₃₈ClNO₂
MDL Number:MFCD02683581