Order Entry
Czech Republic
ContactUsLinkComponent
21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]triene-3,20-dione maleate salt ≥98% (by TLC)
21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]triene-3,20-dione maleate salt ≥98% (by TLC)
Catalog # ENZOBMLFR1000100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
21-[4-(2,6-Di-1-pyrrolidinyl-4-pyrimidinyl)-1-piperazinyl]pregna-1,4,9[11]triene-3,20-dione maleate salt ≥98% (by TLC)
Catalog # ENZOBMLFR1000100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :726.919 g/mol
CAS Number:153190-29-5
Formula:C₄₁H₅₄N₆O₆
MDL Number:MFCD01074991