
6-Ketoprostaglandin-F1α ≥98% (by TLC)
Catalog # ENZOBMLPG0170001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Purity:≥98% (TLC)
- Appearance:White to off-white solid
Specification Test Results
About this item
:
:370.486 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:58962-34-8
Formula:C₂₀H₃₄O₆
MDL Number:MFCD00135222