
Leukotriene B4 ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:71160-24-2
:
Formula:C₂₀H₃₂O₄
MDL Number:MFCD00135630
:335.464 g/mol
Merck Index:14,05455
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Concentration (50 µg size):50 µg/ml
- Concentration (1 mg size):1 mg/ml
- Formulation:Oil dissolved in ethanol.
- Purity:≥99% (HPLC)
- Source:Synthetic
Specification Test Results
Frequently Bought Together