Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Dihydroceramide C8
Dihydroceramide C8 ≥98% (by TLC)
Dihydroceramide C8 ≥98% (by TLC)
ENZOBMLSL1130025
:
Enzo Life Sciences
:
Dihydroceramide C8 ≥98% (by TLC)
ENZOBMLSL1130025
:
Enzo Life Sciences
:
Pk:
25 mg
Seq:
2315
Purity:
≥98% (TLC)
:
:
427.714 g/mol
:
Freezer
:
145774-33-0
:
C₂₆H₅₃NO₃
:
MFCD01320385