0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Manoalide
Manoalide ≥98% (by TLC)
Manoalide ≥98% (by TLC)
Manoalide ≥98% (by TLC)
Catalog # ENZOBMLEI1770001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Manoalide ≥98% (by TLC)
Catalog # ENZOBMLEI1770001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
416.558 g/mol
CAS Number:
75088-80-1
Formula:
C₂₅H₃₆O₅
MDL Number:
MFCD00153826
Product Details