Order Entry
Czech Republic
ContactUsLinkComponent
Phytoceramide C2 ≥98% (by TLC)
Phytoceramide C2 ≥98% (by TLC)
Catalog # ENZOBMLSL1510005
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Phytoceramide C2 ≥98% (by TLC)
Catalog # ENZOBMLSL1510005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :359.551 g/mol
Storage Temperature:Freezer
CAS Number:475995-69-8
Formula:C₂₀H₄₁NO₄
MDL Number:MFCD02686954