Order Entry
Czech Republic
ContactUsLinkComponent
 
Clopamide ≥98% (by HPLC)
Catalog # ENZOBMLCM1120500
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Clopamide ≥98% (by HPLC)
Catalog # ENZOBMLCM1120500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :345.842 g/mol
Storage Temperature:Ambient
CAS Number:636-54-4
Formula:C₁₄H₂₀ClN₃O₃S
MDL Number:MFCD00941495