5-Nitro-2-(3-phenylpropylamino)benzoic acid ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:107254-86-4
:
Formula:C₁₆H₁₆N₂O₄
MDL Number:MFCD00153851
:300.314 g/mol
Storage Temperature:Ambient
Frequently Bought Together