Proadifen hydrochloride ≥95% (by TLC)
Katalogové číslo ENZOBMLEI1051000
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:389.964 g/mol
CAS Number:62-68-0
Formula:C₂₃H₃₁NO₂·HCl
MDL Number:MFCD00055151