Order Entry
Czech Republic
ContactUsLinkComponent
Sphingosine-1-phosphate ≥98% (by TLC)
Irritant
Sphingosine-1-phosphate ≥98% (by TLC)
Catalog # ENZOBMLSL1400001
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
Sphingosine-1-phosphate ≥98% (by TLC)
Catalog # ENZOBMLSL1400001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :379.478 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:26993-30-6
Formula:C₁₈H₃₈NO₅P
MDL Number:MFCD00270077