Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Ethoxyresorufin
7-Ethoxyresorufin ≥98%
7-Ethoxyresorufin ≥98%
7-Ethoxyresorufin ≥98%
Catalog # ENZOBMLEI2620010
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
7-Ethoxyresorufin ≥98%
Catalog # ENZOBMLEI2620010
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
241.246 g/mol
CAS Number:
5725-91-7
Formula:
C₁₄H₁₁NO₃
MDL Number:
MFCD00037661
Product Details