Order Entry
Czech Republic
ContactUsLinkComponent
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98% (by UHPLC)
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98% (by UHPLC)
Katalogové číslo ENZOBMLEI3180100
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98% (by UHPLC)
Katalogové číslo ENZOBMLEI3180100
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :327.90 g/mol
Storage Temperature:Ambient
CAS Number:78628-80-5
Formula:C₂₁H₂₅N
MDL Number:MFCD00145430